CAS RN 2411-83-8
IUPAC Namn:
methyl 2,3-dihydroxybenzoate
Synonymer:
methyl 2,3-dihydroxybenzoate
Molekylvikt (g/mol):
168.15
Molekylär sammansättning:
C8H8O4
InChi Key:
DOAJWTSNTNAEIY-UHFFFAOYSA-N
SMILES:
COC(=O)C1=CC=CC(O)=C1O
1
–
3
av
3
Resultat
1
Methyl 2,3-Dihydroxybenzoate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Methyl 2,3-Dihydroxybenzoate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Methyl 2,3-Dihydroxybenzoate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.