CAS RN 24177-07-9
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
3
av
3
Resultat
1
(2R,5S,10aS,10bS)-Octahydro-10b-hydroxy-5-isobutyl-2-isopropyl-3,6-dioxo-8H-oxazolo[3,2-a]pyrrolo[2,1-c]pyrazine-2-carboxylic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(2R,5S,10aS,10bS)-Octahydro-10b-hydroxy-5-isobutyl-2-isopropyl-3,6-dioxo-8H-oxazolo[3,2-a]pyrrolo[2,1-c]pyrazine-2-carboxylic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(2R,5S,10aS,10bS)-Octahydro-10b-hydroxy-5-isobutyl-2-isopropyl-3,6-dioxo-8H-oxazolo[3,2-a]pyrrolo[2,1-c]pyrazine-2-carboxylic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.