CAS RN 2708218-88-4
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
1
av
1
Resultat
1
Kampanjer är tillgängliga
1H,1H,2H,2H-Perfluorohexanesulfonic acid 13C2 (1,2-13C2) sodium 50 μg/mL in Methanol:Water, Dr. Ehrenstorfer
1H,1H,2H,2H-Perfluorohexanesulfonic acid 13C2 (1,2-13C2) sodium 50 μg/mL in Methanol:Water, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality