CAS RN 2975167-21-4
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
1
av
1
Resultat
1
Tocris Bioscience™ Janelia Fluor 525, NHS ester
Yellow dye (Reactive group: NHS ester). Application: confocal microscopy, super resolution microscopy (SRM) including dSTORM. Suitable for live cell imaging