CAS RN 303176-42-3
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
2
av
2
Resultat
1
(2S,AlphaR)-Alpha-(Aminomethyl)-6-fluoro-3,4-dihydro-2H-1-benzopyran-2-methanol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(2S,AlphaR)-Alpha-(Aminomethyl)-6-fluoro-3,4-dihydro-2H-1-benzopyran-2-methanol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.