CAS RN 3348-03-6
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
1
av
1
Resultat
1
MedChemExpress 6-CFDA
MedChemExpress 6-CFDA is a fluorescent probe that can be used in flow cytometry and fluorescence microscopy. 6-CFDA also can be applied in intravital multiphoton microscopy to research hepatobiliary metabolism in animals.