CAS RN 346-55-4
IUPAC Namn:
4-chloro-7-(trifluoromethyl)quinoline
Synonymer:
4-chloro-7-(trifluoromethyl)quinoline
Molekylvikt (g/mol):
231.6
Molekylär sammansättning:
C10H5ClF3N
InChi Key:
LLRQVSZVVAKRJA-UHFFFAOYSA-N
SMILES:
FC(F)(F)C1=CC=C2C(Cl)=CC=NC2=C1
1
–
1
av
1
Resultat
1
4-Chloro-7-(trifluoromethyl)quinoline, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.