CAS RN 34885-03-5
IUPAC Namn:
(4-methylcyclohexyl)methanol
Synonymer:
4-methylcyclohexanemethanol
Molekylvikt (g/mol):
128.22
Molekylär sammansättning:
C8H16O
InChi Key:
OSINZLLLLCUKJH-UHFFFAOYSA-N
SMILES:
CC1CCC(CO)CC1
1
–
3
av
3
Resultat
1
4-Methylcyclohexanemethanol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
4-Methylcyclohexanemethanol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
4-Methylcyclohexanemethanol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.