CAS RN 3516-95-8
IUPAC Namn:
1-(2-hydroxyphenyl)-3-phenylpropan-1-one
Synonymer:
1-(2-hydroxyphenyl)-3-phenylpropan-1-one
Molekylvikt (g/mol):
226.28
Molekylär sammansättning:
C15H14O2
InChi Key:
JCPGMXJLFWGRMZ-UHFFFAOYSA-N
SMILES:
OC1=CC=CC=C1C(=O)CCC1=CC=CC=C1
1
–
3
av
3
Resultat
1
Propafenone EP Impurity A, Mikromol
Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
2’-Hydroxy-3-phenylpropiophenone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
2’-Hydroxy-3-phenylpropiophenone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.