CAS RN 362049-46-5
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
2
av
2
Resultat
1
2,3,5-Trimethylphenol-d11, CDN
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
2
2,3,5-Trimethylphenol-d11, TRC
CAS: 362049-46-5 Molekylformel: C9 D11 H O Molekylvikt (g/mol): 147.2588 Synonym: Phen-2,4-d2-ol, 3,5,6-tri(methyl-d3)- (9CI),2,3,5-Trimethylphenol-d11,2,3,5-Trimethylphenol-D11,2,3,5-Tri(methyl-d3)phenol-4,6-d2,3,5,6-Tri(methyl-d3)phenol-2,4-d2 IUPAC-namn: 2,4-dideuterio-3,5,6-tris(trideuteriomethyl)phenol LEDER: [2H]c1c(O)c(c(c([2H])c1C([2H])([2H])[2H])C([2H])([2H])[2H])C([2H])([2H])[2H]