CAS RN 382-28-5
IUPAC Namn:
2,2,3,3,5,5,6,6-octafluoro-4-(trifluoromethyl)morpholine
Synonymer:
perfluoro-4-methylmorpholine
Molekylvikt (g/mol):
299.04
Molekylär sammansättning:
C5F11NO
InChi Key:
PQMAKJUXOOVROI-UHFFFAOYSA-N
SMILES:
FC(F)(F)N1C(F)(F)C(F)(F)OC(F)(F)C1(F)F
1
–
3
av
3
Resultat
1
Perfluoro-N-methylmorpholine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Perfluoro-N-methylmorpholine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Perfluoro-N-methylmorpholine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.