CAS RN 39878-65-4
IUPAC Namn:
2-amino-3-(4-methoxyphenyl)propanoic acid
Synonymer:
2-amino-3-(4-methoxyphenyl)propanoic acid
Molekylvikt (g/mol):
195.22
Molekylär sammansättning:
C10H13NO3
InChi Key:
GEYBMYRBIABFTA-UHFFFAOYNA-N
SMILES:
COC1=CC=C(CC(N)C(O)=O)C=C1
1
–
3
av
3
Resultat
1
(R)-2-Amino-3-(4-methoxyphenyl)propanoic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(R)-2-Amino-3-(4-methoxyphenyl)propanoic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(R)-2-Amino-3-(4-methoxyphenyl)propanoic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.