CAS RN 4816-80-2
IUPAC Namn:
2-(4-methylbenzenesulfonamido)pentanedioic acid
Synonymer:
2-(4-methylbenzenesulfonamido)pentanedioic acid
Molekylvikt (g/mol):
301.31
Molekylär sammansättning:
C12H15NO6S
InChi Key:
KKOZKXBAPIYWAT-UHFFFAOYNA-N
SMILES:
CC1=CC=C(C=C1)S(=O)(=O)NC(CCC(O)=O)C(O)=O
1
–
1
av
1
Resultat
1
N-Tosyl-L-glutamic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.