CAS RN 5026-42-6
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
3
av
3
Resultat
1
5,10-Dihydro-5-methyl-11H-dibenzo[b,e][1,4]diazepin-11-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
5,10-Dihydro-5-methyl-11H-dibenzo[b,e][1,4]diazepin-11-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
5,10-Dihydro-5-methyl-11H-dibenzo[b,e][1,4]diazepin-11-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.