CAS RN 51072-36-7
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
3
av
3
Resultat
1
1,2,3,4,5,6,7,8-Octahydro-1-[(4-methoxyphenyl)methyl]-isoquinoline, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
1,2,3,4,5,6,7,8-Octahydro-1-[(4-methoxyphenyl)methyl]-isoquinoline, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
1,2,3,4,5,6,7,8-Octahydro-1-[(4-methoxyphenyl)methyl]-isoquinoline, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.