CAS RN 5175-83-7
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
1
av
1
Resultat
1
Tris(2,4,6-Tribromophenoxy)-bismuthine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.