CAS RN 5580-79-0
IUPAC Namn:
1,2,3,4-tetrafluoro-5-nitrobenzene
Synonymer:
1,2,3,4-tetrafluoro-5-nitrobenzene
Molekylvikt (g/mol):
195.07
Molekylär sammansättning:
C6HF4NO2
InChi Key:
MKMDVNZEIQDZEP-UHFFFAOYSA-N
SMILES:
[O-][N+](=O)C1=CC(F)=C(F)C(F)=C1F
1
–
3
av
3
Resultat
1
2,3,4,5-Tetrafluoronitrobenzene, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2,3,4,5-Tetrafluoronitrobenzene, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
2,3,4,5-Tetrafluoronitrobenzene, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.