CAS RN 56396-35-1
IUPAC Namn:
(2E)-2-cyano-3-(1-phenyl-1H-indol-3-yl)prop-2-enoic acid
Synonymer:
(2E)-2-cyano-3-(1-phenylindol-3-yl)prop-2-enoic acid
Molekylvikt (g/mol):
288.31
Molekylär sammansättning:
C18H12N2O2
InChi Key:
BIZNHCWFGNKBBZ-JLHYYAGUSA-N
SMILES:
OC(=O)C(=C\C1=CN(C2=CC=CC=C12)C1=CC=CC=C1)\C#N
1
–
3
av
3
Resultat
1
UK-5099, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Tocris Bioscience™ UK 5099
CAS: 56396-35-1 Molekylformel: C18H11N2O2-
3
UK-5099, MedChemExpress
MedChemExpress UK-5099 (PF-1005023) is a potent inhibitor of the mitochondrial pyruvate carrier (MPC). UK-5099 (PF-1005023) inhibits pyruvate-dependent O2 consumption with an IC50 of 50 nM.