CAS RN 57704-36-6
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
3
av
3
Resultat
1
D-threo-1-(4-Aminophenyl)-2-dichloroacetylamino-1,3-propanediol Hydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
D-threo-1-(4-Aminophenyl)-2-dichloroacetylamino-1,3-propanediol Hydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
D-threo-1-(4-Aminophenyl)-2-dichloroacetylamino-1,3-propanediol Hydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.