CAS RN 589-34-4
IUPAC Namn:
3-methylhexane
Synonymer:
3-methylhexane
Molekylvikt (g/mol):
100.21
Molekylär sammansättning:
C7H16
InChi Key:
VLJXXKKOSFGPHI-UHFFFAOYNA-N
SMILES:
CCCC(C)CC
1
–
2
av
2
Resultat
1
3-Methylhexane, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality