CAS RN 59467-86-6
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
3
av
3
Resultat
1
8-Chloro-1-methyl-6-phenyl-4H-imidazo[1,5-a][1,4]benzodiazepine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
8-Chloro-1-methyl-6-phenyl-4H-imidazo[1,5-a][1,4]benzodiazepine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
8-Chloro-1-methyl-6-phenyl-4H-imidazo[1,5-a][1,4]benzodiazepine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.