CAS RN 617-79-8
IUPAC Namn:
3-(aminomethyl)pentane
Synonymer:
2-ethylbutylamine
Molekylvikt (g/mol):
101.19
Molekylär sammansättning:
C6H15N
InChi Key:
MGWAGIQQTULHGU-UHFFFAOYSA-N
SMILES:
CCC(CC)CN
1
–
3
av
3
Resultat
1
2-Ethylbutylamine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2-Ethylbutylamine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
2-Ethylbutylamine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.