CAS RN 627-76-9
IUPAC Namn:
2-aminoheptanedioic acid
Synonymer:
2-aminopimelic acid
Molekylvikt (g/mol):
175.18
Molekylär sammansättning:
C7H13NO4
InChi Key:
JUQLUIFNNFIIKC-UHFFFAOYNA-N
SMILES:
NC(CCCCC(O)=O)C(O)=O
1
–
2
av
2
Resultat
1
D,L-Alpha-Aminopimelic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
D,L-Alpha-Aminopimelic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.