CAS RN 629-80-1
IUPAC Namn:
hexadecanal
Synonymer:
hexadecanal palmitoyl aldehyde PLM PLY N-hexadecanal
Molekylvikt (g/mol):
240.43
Molekylär sammansättning:
C16H32O
InChi Key:
NIOYUNMRJMEDGI-UHFFFAOYSA-N
SMILES:
CCCCCCCCCCCCCCCC=O
1
–
3
av
3
Resultat
1
Hexadecanal, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Hexadecanal, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Hexadecanal, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.