CAS RN 64963-01-5
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
3
av
3
Resultat
1
MedChemExpress [D-Ala2]leucine-enkephalin
MedChemExpress [D-Ala2]leucine-enkephalin, a delta opioid agonist, is a degradation resistant long-acting Leu-enkephalin.
2
MedChemExpress [D-Ala2]leucine-enkephalin
MedChemExpress [D-Ala2]leucine-enkephalin, a delta opioid agonist, is a degradation resistant long-acting Leu-enkephalin.
3
MedChemExpress [D-Ala2]leucine-enkephalin
MedChemExpress [D-Ala2]leucine-enkephalin, a delta opioid agonist, is a degradation resistant long-acting Leu-enkephalin.