CAS RN 714964-61-1
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
3
av
3
Resultat
1
N-tert-Boc-L-alanine-D4, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
L-Alanine-2,3,3,3-d4-N-t-BOC, CDN
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
3
N-tert-Boc-L-alanine-D4, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.