CAS RN 72522-86-2
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
3
av
3
Resultat
1
(-)-8,9-Dihydroxy-8,9-dihydrobenz[a]anthracene, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(-)-8,9-Dihydroxy-8,9-dihydrobenz[a]anthracene, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(-)-8,9-Dihydroxy-8,9-dihydrobenz[a]anthracene, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.