CAS RN 786-19-6
IUPAC Namn:
O,O-diethyl ({[(4-chlorophenyl)sulfanyl]methyl}sulfanyl)phosphonothioate
Synonymer:
carbophenothion 卡波硫磷 carbophenthion
Molekylvikt (g/mol):
342.85
Molekylär sammansättning:
C11H16ClO2PS3
InChi Key:
VEDTXTNSFWUXGQ-UHFFFAOYSA-N
SMILES:
CCOP(=S)(OCC)SCSC1=CC=C(Cl)C=C1
1
–
3
av
3
Resultat
1
Carbophenothion, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Carbophenothion, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.