CAS RN 887406-71-5
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
3
av
3
Resultat
1
4-(Maleimidomethyl)cyclohexane-1-carbonyl-1-(tert-butyl)carbazate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
4-(Maleimidomethyl)cyclohexane-1-carbonyl-1-(tert-butyl)carbazate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
4-(Maleimidomethyl)cyclohexane-1-carbonyl-1-(tert-butyl)carbazate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.