CAS RN 90057-09-3
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
3
av
3
Resultat
1
2,6-Dimethoxy-7-deazapurine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2,6-Dimethoxy-7-deazapurine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
2,6-Dimethoxy-7-deazapurine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.