CAS RN 917879-39-1
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
1
av
1
Resultat
1
MK-2461, MedChemExpress
MedChemExpress MK-2461 is a novel ATP-competitive multitargeted inhibitor of activated c-Met with a mean IC50 of 2.5 nM.