CAS RN 924-42-5
IUPAC Namn:
N-(hydroxymethyl)prop-2-enamide
Synonymer:
N-(hydroxymethyl)acrylamide
Molekylvikt (g/mol):
101.11
Molekylär sammansättning:
C4H7NO2
InChi Key:
CNCOEDDPFOAUMB-UHFFFAOYSA-N
SMILES:
OCNC(=O)C=C
1
–
2
av
2
Resultat
1
N-(Hydroxymethyl)acrylamide 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
2
N-(Hydroxymethyl)acrylamide, TRC
CAS: 924-42-5 Molekylformel: C4 H7 N O2 Molekylvikt (g/mol): 101.1 Synonym: Acrylamide, N-(hydroxymethyl)- (6CI,8CI),N-(Hydroxymethyl)-2-propenamide,Cylink NMA,MH 100,MH 100 (amide),Monomethylolacrylamide,N-(Hydroxymethyl)acrylamide,N-MAM,N-MAM P,N-Methanolacrylamide,N-Methylolacrylamide,N-NBM,NMA 60,NSC 553,Rocagil BT,U-Ramin T 80 IUPAC-namn: N-(hydroxymethyl)prop-2-enamide LEDER: OCNC(=O)C=C