Läs mer
N-(Hydroxymethyl)acrylamide, TRC
Produktkod. 30136877
Change view
Kvantitet:
250 mg
500 mg
2.5 g
Förpackningsstorlek:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30136877 500 mg -
30132772 250 mg -
30010107 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30136877 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH511405500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 924-42-5
Molekylformel C4 H7 N O2
Molekylvikt (g/mol) 101.1
Synonym Acrylamide, N-(hydroxymethyl)- (6CI,8CI), N-(Hydroxymethyl)-2-propenamide, Cylink NMA, MH 100, MH 100 (amide), Monomethylolacrylamide, N-(Hydroxymethyl)acrylamide, N-MAM, N-MAM P, N-Methanolacrylamide, N-Methylolacrylamide, N-NBM, NMA 60, NSC 553, Rocagil BT, U-Ramin T 80
IUPAC-namn N-(hydroxymethyl)prop-2-enamide
LEDER OCNC(=O)C=C
Specifikationer
Kemiskt namn eller material N-(Hydroxymethyl)acrylamide
Kvantitet 500 mg
InChI formel InChI=1S/C4H7NO2/c1-2-4(7)5-3-6/h2,6H,1,3H2,(H,5,7)
Formel vikt 101.0477