CAS RN 936021-98-6
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
2
av
2
Resultat
1
1,2-Benzenedicarboxylic Acid 1-(7-Hydroxy-4-methyloctyl) Ester (Mixture of Diastereomers), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
1,2-Benzenedicarboxylic Acid 1-(7-Hydroxy-4-methyloctyl) Ester (Mixture of Diastereomers), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.