CAS RN 96333-59-4
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
3
av
3
Resultat
1
5,7-Bis-(benzyloxy)-2-(4-(benzyloxy)phenyl)-4H-chromen-4-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
5,7-Bis-(benzyloxy)-2-(4-(benzyloxy)phenyl)-4H-chromen-4-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
5,7-Bis-(benzyloxy)-2-(4-(benzyloxy)phenyl)-4H-chromen-4-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.