CAS RN 986-96-9
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
3
av
3
Resultat
1
3,17-Dihydroxy-6-methyl-pregna-3,5-dien-20-one Diacetate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
3,17-Dihydroxy-6-methyl-pregna-3,5-dien-20-one Diacetate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
3,17-Dihydroxy-6-methyl-pregna-3,5-dien-20-one Diacetate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.