missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Respiratory Syncytial Virus Glycoprotein G Antibody (133) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50411-0.025ml
This item is not returnable.
View return policy
Description
Respiratory Syncytial Virus Glycoprotein G Monoclonal antibody specifically detects Respiratory Syncytial Virus Glycoprotein G in Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Specifications
| Respiratory Syncytial Virus Glycoprotein G | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence | |
| 133 | |
| Unconjugated | |
| PBS | |
| Human RSV strain A2 infected HeLa cells | |
| 0.025 mL | |
| Virology Bacteria and Parasites | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction