missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aurora B Antibody (3F11), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50039
This item is not returnable.
View return policy
Description
Aurora B Monoclonal antibody specifically detects Aurora B in Human, Mouse, Rat, Bovine, Equine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Proximity Ligation Assay
Specifications
Aurora B | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:1000, Immunohistochemistry 1:1000, Immunocytochemistry/ Immunofluorescence 1:1000, Proximity Ligation Assay | |
Q96GD4, Q96GD4 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Bovine, Equine | |
Purified |
Western Blot, Immunohistochemistry, Immunofluorescence, Proximity Ligation Assay | |
3F11 | |
Unconjugated | |
50% PBS, 50% glycerol | |
AIK2EC 2.7.11.1, AIM1Aik2, AIM-1STK-1, ARK2ARK-2, AurB, aurkb-sv2, Aurora- and IPL1-like midbody-associated protein 1, aurora kinase BSerine/threonine-protein kinase aurora-B, aurora kinase B-Sv1, aurora kinase B-Sv2, Aurora/IPL1-related kinase 2, aurora-1, aurora-B, Aurora-related kinase 2, EC 2.7.11, IPL1, serine/threonine kinase 12, serine/threonine-protein kinase 12, STK12aurkb-sv1, STK5 | |
Full length recombinant human aurora B protein expressed in and purified from E. coli. | |
0.1 mL | |
Breast Cancer, Cancer, Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, DNA Repair, Mitotic Regulators, Stem Cell Markers | |
9212 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Aurora B Antibody (3F11), Novus Biologicals™ > 0.1 mL, Unconjugated
Spot an opportunity for improvement?Share a Content Correction