missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDC42 Antibody (M152), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-23212
This item is not returnable.
View return policy
Description
CDC42 Monoclonal antibody specifically detects CDC42 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence
Specifications
CDC42 | |
Monoclonal | |
Unconjugated | |
PBS, 1 mg/ml BSA and 50% glycerol | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunofluorescence | |
M152 | |
Western Blot 1:500, Immunocytochemistry/Immunofluorescence 1:50 | |
CDC42Hs, cell division control protein 42 homolog, cell division cycle 42 (GTP binding protein, 25kDa), cell division cycle 42 (GTP-binding protein, 25kD), dJ224A6.1.1 (cell division cycle 42 (GTP-binding protein, 25kD)), dJ224A6.1.2 (cell division cycle 42 (GTP-binding protein, 25kD)), G25K, G25K GTP-binding protein, growth-regulating protein, GTP-binding protein, 25kD, small GTP binding protein CDC42 | |
Clone (M152) was generated from a rat recombinant Cdc42 protein. This sequence is highly conserved in human and mouse Cdc42. [UniProt# Q8CFN2] | |
0.1 mL | |
Breast Cancer, Cancer, Phospho Specific, Signal Transduction | |
998 | |
Store at -20°C. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CDC42 Antibody (M152), Novus Biologicals™ > 0.1 mL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction