Learn More
OASIS/CREB3L1 Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-00039PCP
This item is not returnable.
View return policy
Description
OASIS/CREB3L1 Polyclonal antibody specifically detects OASIS/CREB3L1 in Human samples. It is validated for ELISA
Specifications
OASIS/CREB3L1 | |
Polyclonal | |
PBS | |
Rabbit | |
0.1 mL | |
Store at 4C in the dark. |
ELISA | |
PerCP | |
BBF-2 homolog, cAMP responsive element binding protein 3-like 1, cAMP-responsive element-binding protein 3-like protein 1, OASIScyclic AMP-responsive element-binding protein 3-like protein 1, old astrocyte specifically induced substance, Old astrocyte specifically-induced substance | |
Produced in rabbits immunized with purified, recombinant Human OASIS/CREB3L1 (Uniprot#: Q96BA8-1; Glu396- Ser519) | |
90993 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
OASIS/CREB3L1 Antibody, PerCP, Novus Biologicals™ > 0.1 mL; PerCP
Spot an opportunity for improvement?Share a Content Correction