missing translation for 'onlineSavingsMsg'
Learn More
Learn More
E-Tag Epitope Tag Antibody [PE], Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-43762PE
This item is not returnable.
View return policy
Description
E-Tag Epitope Tag Polyclonal antibody specifically detects E-Tag Epitope Tag in Epitope Tag samples. It is validated for Immunoprecipitation,Western Blot
Specifications
E-Tag Epitope Tag | |
Polyclonal | |
Western Blot, Immunoprecipitation | |
E epitope tag, GAPVPYPDPLEPR epitope tag, GAPVPYPDPLEPR tag | |
Produced in rabbits immunized with a synthetic peptide corresponding to the E-Tag Epitope Tag sequence (GAPVPYPDPLEPR) | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. | |
IgG |
Immunoprecipitation, Western Blot | |
PE | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Tag | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
E-Tag Epitope Tag Antibody [PE], Novus Biologicals™ > 0.1 mL; PE
Spot an opportunity for improvement?Share a Content Correction