Learn More
Novus Biologicals™ Syntaxin Binding Protein 4 293T Cell Transient Overexpression Lysate
Brand:  Novus Biologicals™ H00252983-T01
Product Code. 18486419
  • 4495.00 SEK / 0.10mL
Specifications
0.1 mL
Endocrinology, Signal Transduction
FLJ16496, MGC50337, ST x 4-interacting protein, SynipMGC149829, syntaxin 4 interacting protein, Syntaxin 4-interacting protein, syntaxin binding protein 4, syntaxin-binding protein 4
Store at -80°C. Avoid freeze/thaw cycles.
Syntaxin Binding Protein 4
STXBP4
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Novus Biologicals™ Syntaxin Binding Protein 4 293T Cell Transient Overexpression Lysate > 0.1 mL