Läs mer
[1',2',3',4',5'-13C5]Uridine, TRC
Produktkod. 30456379
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30456379 10 mg
30451774 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30456379 Leverantör Toronto Research Chemicals Leverantörsnummer TRCU82991710MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 159496-16-9
Molekylformel 13C5 C4 H12 N2 O6
Molekylvikt (g/mol) 249.16
Synonym Uridine-1',2',3',4',5'-13C5 (9CI), [1',2',3',4',5'-13C5]Uridine, 1-(β-D-Ribofuranosyl-1,2,3,4,5-13C5)-2,4(1H,3H)-pyrimidinedione, 1-(β-D-Ribofuranosyl-1,2,3,4,5-13C5)uracil
IUPAC-namn 1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxy(1^{13}C)methyl)(2,3,4,5-^{13}C_{4})oxolan-2-yl]pyrimidine-2,4-dione
LEDER O[13CH2][13C@H]1O[13C@H]([13C@H](O)[13C@@H]1O)N2C=CC(=O)NC2=O
Specifikationer
Kemiskt namn eller material Uridine-1',2',3',4',5'-13C5
Kvantitet 10 mg
InChI formel InChI=1S/C9H12N2O6/c12-3-4-6(14)7(15)8(17-4)11-2-1-5(13)10-9(11)16/h1-2,4,6-8,12,14-15H,3H2,(H,10,13,16)/t4-,6-,7-,8-/m1/s1/i3+1,4+1,6+1,7+1,8+1
Formel vikt 249.0863