Läs mer
1,3,5-Trichloro-2-methylbenzene, TRC
Produktkod. 17930985
Change view
Kvantitet:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17930985 25 mg
17942005 50 mg
17902195 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17930985 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT77436325MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 23749-65-7
Molekylformel C7 H5 Cl3
Molekylvikt (g/mol) 195.47
Synonym 1,3,5-Trichloro-2-methylbenzene (ACI), Toluene, 2,4,6-trichloro- (8CI), 2,4,6-Trichlorotoluene, 1,3,5-Trichloro-2-methylbenzene (ACI), Toluene, 2,4,6-trichloro- (8CI), 2,4,6-Trichlorotoluene, 2,4,6-Trichloro-toluene;
IUPAC-namn 1,3,5-trichloro-2-methylbenzene
LEDER Cc1c(Cl)cc(Cl)cc1Cl
Specifikationer
Kemiskt namn eller material 1,3,5-Trichloro-2-methylbenzene
Kvantitet 25 mg
Anteckningar om renhetsgrad GC
InChI formel InChI=1S/C7H5Cl3/c1-4-6(9)2-5(8)3-7(4)10/h2-3H,1H3
Formel vikt 193.946
Procent renhet >95%