Learn More
Trajan Scientific and Medical™ 1/4 in. Graphite/Vespel Fittings
Provide leak-free system connections. Trajan Scientific and Medical 1/4 in. Graphite/Vespel Fittings leads to improved chromatography for non-MS systems.
Brand:  Trajan Scientific and Medical™ 0726660
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15288893
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • Composite of graphite and Vespel
  • Mechanically robust, hard material, long lifetime
  • Forms a strong grip with the capillary column
Specifications
Specifications
9.5 x 7.4 mm
9.5 mm
7.4 mm
0.5 mm
Bore size: 0.5 mm
15% Graphite, 85% Vespel
Graphite Vespel Reducing Ferrule 1/4 in. Fitting 0.5mm x 9.5mm x 7.4mm