Learn More
1-(bromomethyl)-2-phenoxybenzene, 90%, Thermo Scientific™ Product Code.: 10029142
1-(bromomethyl)-2-phenoxybenzene, 90%, Thermo Scientific™
Product Code. p-4510711
250 mg, 250mg
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
Product Code. 10029142
Brand: Thermo Scientific Maybridge CC49908CB
Request Bulk or Custom Format
Chemical Identifiers
CAS 82657-72-5
Molecular Formula C13H11BrO
Molecular Weight (g/mol) 263.134
InChI Key YQRIQBOWLXRKKG-UHFFFAOYSA-N
Synonym 1-bromomethyl-2-phenoxybenzene, 2-phenoxybenzyl bromide, benzene, bromomethylphenoxy, acmc-20eqyy, 2-bromomethylphenyl phenyl ether, 1-bromomethyl-2-phenoxy-benzene, 2-bromomethyl-1-phenoxybenzene, 1-2-bromomethyl phenoxy benzene, 1-bromomethyl-2-phenyloxy benzene, benzene, 1-bromomethyl-2-phenoxy
PubChem CID 22675469
IUPAC Name 1-(bromomethyl)-2-phenoxybenzene
SMILES C1=CC=C(C=C1)OC2=CC=CC=C2CBr
Specifications
Assay Percent Range 97%
Quantity 250 mg
Formula Weight 263.13
Percent Purity 97%
Chemical Name or Material 1-(Bromomethyl)-2-phenoxybenzene
Product Content Correction
Product Title
1-(bromomethyl)-2-phenoxybenzene, 90%, Thermo Scientific™ >