missing translation for 'onlineSavingsMsg'
Läs mer

1-Chloro-4-(trifluoromethoxy)benzene, 98%, Thermo Scientific™

Produktkod. 11489417
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
2 g
10 g
Förpackningsstorlek:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet Förpackningsstorlek
11489417 2 g 2g
11499417 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 11489417 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer B21437.04

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 461-81-4
Molekylformel C7H4ClF3O
Molekylvikt (g/mol) 196.553
MDL-nummer MFCD00040953
InChI-nyckel SELFZOLQRDPBKC-UHFFFAOYSA-N
Synonym 4-trifluoromethoxy chlorobenzene, 1-chloro-4-trifluoromethoxy benzene, 4-chloro trifluoromethoxy benzene, p-chlorotrifluoromethoxybenzene, benzene, 1-chloro-4-trifluoromethoxy, 4-chloro-alpha,alpha,alpha-trifluoroanisole, 4-chloro-1-trifluoromethoxy benzene, pubchem4348, acmc-1aj10, ksc494s4h
PubChem CID 2777273
IUPAC-namn 1-chloro-4-(trifluoromethoxy)benzene
LEDER C1=CC(=CC=C1OC(F)(F)F)Cl

Specifications

Kemiskt namn eller material 1-Chloro-4-(trifluoromethoxy)benzene
Densitet 1.37
Kvantitet 2 g
Kokpunkt 142°C to 143°C
Formel vikt 196.56
Procent renhet 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.