missing translation for 'onlineSavingsMsg'
Learn More
1-Chlorobutane CHROMASOLV™, for HPLC, ≥99.8%, Honeywell Riedel-de Haën™ Product Code.: 15694720

1-Chlorobutane CHROMASOLV™, for HPLC, ≥99.8%, Honeywell Riedel-de Haën™

Product Code. 15694720
1 L, 1 L
Click to view available options
Quantity:
1 L
2 L
2.5 L
Unit Size:
1 L
2 L
2.50 L
This item is not returnable. View return policy

Product Code. 15694720

Brand: Honeywell-Chromasolv 349581L

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CHROMASOLV™, for HPLC,≥99.8%

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-69-3
Molecular Formula C4H9Cl
Molecular Weight (g/mol) 92.57
MDL Number MFCD00001009
InChI Key VFWCMGCRMGJXDK-UHFFFAOYSA-N
Synonym butyl chloride, n-butyl chloride, butane, 1-chloro, chlorure de butyle, nbc wormer, n-propylcarbinyl chloride, sure shot, butane, chloro, chlorobutane, n-chlorobutane
PubChem CID 8005
IUPAC Name 1-chlorobutane
SMILES CCCCCl

Specifications

Melting Point -123°C
Density 0.886g/mL at 25°C (lit.)
Boiling Point 77°C to 78°C
Linear Formula CH3(CH2)3Cl
Refractive Index n20/D 1.402
Quantity 1 L
Beilstein 1730909
Formula Weight 92.57g/mol
Vapor Pressure 80.1mmHg (78.4°C)
Percent Purity ≥99.8%
Grade CHROMASOLV™
Chemical Name or Material 1-Chlorobutane, for HPLC
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Chlorobutane
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Aspiration toxicity Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Chlorobutane CHROMASOLV™, for HPLC, ≥99.8%, Honeywell Riedel-de Haën™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.