Learn More
BRAND™ 1 D Filter Crucible
Filter Crucible constructed of clear glass for visibility.
Brand:  BRAND™ 458023
3182.49 SEK valid until 2024-12-31
Use promo code "23264" to get your promotional price.
Additional Details : Weight : 3.10000kg
 View more versions of this product
Product Code. 15202449
  • 3565.00 SEK / Pack of 10
Estimated Shipment: 16-05-2024
to see stock.
Add to basket
Description
Description
  • Constructed of Borosilicate Glass 3.3
Specifications
Specifications
3 cm
30 mL
3
Borosilicate Glass
Crucible