Learn More
Invitrogen CELLSENSOR(R) ARE-BLA HEP G2 CELL LINE 1ml
Brand:  Invitrogen K1208
CELLSENSOR(R) ARE-BLA HEP G2 CELL LINE 1ml
Product Code. 10024572
  • 159500.00 SEK / 1mL
For Research Use Only. All usage must comply with product instructions.
Product Title
Invitrogen CELLSENSOR(R) ARE-BLA HEP G2 CELL LINE 1ml >